Please login to your online account to display your discounted pricing

L-(+)-Arginine Monohydrochloride, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™


High quality biochemicals for research use

Quantity 100g
Packaging Poly Bottle

Manufacturer: avantor performance  B57705

 View more versions of this product

Catalog No. 02003145

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 210.67
CAS 1119-34-2
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material L-(+)-Arginine Monohydrochloride
Molecular Formula NH2C(:NH)NH(CH2)3CH(NH2)COOH·HCl
Quantity 100g
Packaging Poly Bottle
Specific Gravity 1L = 1.42kg

Specifications

Chemical Name or Material L-(+)-Arginine Monohydrochloride
CAS 1119-34-2
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula NH2C(:NH)NH(CH2)3CH(NH2)COOH·HCl
Formula Weight 210.67