Please login to your online account to display your discounted pricing

L-(+)-Arginine Monohydrochloride, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$239.72 - $1,615.26

Chemical Identifiers

Chemical Name or Material L-(+)-Arginine Monohydrochloride
CAS 1119-34-2
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula NH2C(:NH)NH(CH2)3CH(NH2)COOH·HCl
Formula Weight 210.67
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003145

 
avantor performance
B57705
Poly Bottle 100g Each for $239.72

02003147

 
avantor performance
B57706
Poly Bottle 1kg Each for $1,615.26
Description & Specifications

Specifications

Formula Weight 210.67
CAS 1119-34-2
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material L-(+)-Arginine Monohydrochloride
Molecular Formula NH2C(:NH)NH(CH2)3CH(NH2)COOH·HCl
Quantity 100g
Packaging Poly Bottle
Specific Gravity 1L = 1.42kg