Please login to your online account to display your discounted pricing

L(+)-Arginine Hydrochloride, 98+%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  105001000

 View more versions of this product

Catalog No. AC105001000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 210.67
Quantity 100g
Color White
Assay 98.0 to 102.0%
Specific Rotation +21.4° to +23.6° (20°C, 589nm) (c=8, 6N HCl)
Sulfate 300ppm max.
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Chemical Name or Material L(+)-Arginine hydrochloride
Assay Percent Range 98+%
Other Amino Acids 0.5% max. (TLC)
Sulfated Ash 0.1% max.
Heavy Metals (as Pb) 20ppm max.
Packaging Plastic bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Molecular Formula C6H14N4O2·HCl
MDL Number MFCD00064550
CAS 1119-34-2

Specifications

Chemical Name or Material L(+)-Arginine hydrochloride
Assay 98.0 to 102.0%
Assay Percent Range 98+%
Molecular Formula C6H14N4O2·HCl
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Formula Weight 210.67
MDL Number MFCD00064550