missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents L-Arginine, Free Base, Fisher BioReagents
Supplier: Fisher BioReagents BP2505500
Description
L-Arginine is an essential amino acid used in biochemical research applications.Specifications
223°C | |
74-79-3 | |
White | |
Pass Test | |
Conforms to reference | |
0.3% max. | |
Poly Bottle | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid | |
174.20 | |
CHEBI:16467 | |
≥99% | |
Solid |
L-Arginine, Free Base | |
11.4 | |
≥99 % | |
C6H14N4O2 | |
MFCD00002635 | |
15, 770 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
NC(CCCN=C(N)N)C(O)=O | |
+26.3 to +27.7° (+ or -) | |
6322 | |
174.12 | |
500 g |
Chemical Identifiers
74-79-3 | |
174.20 | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
6322 | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid |
C6H14N4O2 | |
MFCD00002635 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
CHEBI:16467 | |
NC(CCCN=C(N)N)C(O)=O |
Safety and Handling

Recommended Storage : RT