missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents L-Arginine, Free Base, Fisher BioReagents

Supplier:  Fisher BioReagents BP25051

 View more versions of this product

Catalog No. BP25051




Description

Description

L-Arginine is an essential amino acid used in biochemical research applications.
Specifications

Specifications

223°C
74-79-3
White
Pass Test
Conforms to reference
0.3% max.
Poly Bottle
ODKSFYDXXFIFQN-UHFFFAOYNA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
174.20
CHEBI:16467
≥99%
Solid
L-Arginine, Free Base
11.4
≥99 %
C6H14N4O2
MFCD00002635
15, 770
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
NC(CCCN=C(N)N)C(O)=O
+26.3 to +27.7° (+ or -)
6322
174.12
1 kg

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
6322
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
CHEBI:16467
NC(CCCN=C(N)N)C(O)=O
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions