missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Arginine, 98+%, Alfa Aesar

Manufacturer:  Alfa Aesar A157380E

 View more versions of this product

Catalog No. AAA157380E



Description

Description

Solubility
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

L-Arginine
98+%
MFCD00002635
14,780
ODKSFYDXXFIFQN-BYPYZUCNSA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
6322
174.2
226°C (decomposition)
+26° (c=1.6 in 6N HCl)
74-79-3
C6H14N4O2
1725413
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, l-arginine, arginina, arginine, l
C(CC(C(=O)O)N)CN=C(N)N
174.204
CHEBI:16467
≥98%
2500g

Chemical Identifiers

74-79-3
174.204
ODKSFYDXXFIFQN-BYPYZUCNSA-N
6322
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, l-arginine, arginina, arginine, l
CHEBI:16467
C(CC(C(=O)O)N)CN=C(N)N
Safety and Handling

Safety and Handling

RTECSNumber : CF1934200

TSCA : TSCA

storageNote1 : Air sensitive

Documents