missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Arginine 4-nitroanilide dihydrochloride, 95%

$40.09 - $578.30

Chemical Identifiers

CAS 40127-11-5
Molecular Formula C12H20Cl2N6O3
Molecular Weight (g/mol) 367.231
MDL Number MFCD00058249
InChI Key FBVMDLFQVOFFHS-XRIOVQLTSA-N
Synonym h-arg-pna.2hcl, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide dihydrochloride, l-arginine p-nitroanilide dihydrochloride, arginine p-nitroanilide dihydrochloride, 2s-2-amino-5-carbamimidamido-n-4-nitrophenyl pentanamide dihydrochloride, h-arg-pna 2hcl, h-arg-pna.dihydrochloride, h-arg-pna inverted exclamation mark currency2hcl, n-4-nitrophenyl argininamide dihydrochloride, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide 2hcl
PubChem CID 22811858
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)-N-(4-nitrophenyl)pentanamide;dihydrochloride
SMILES C1=CC(=CC=C1NC(=O)C(CCCN=C(N)N)N)[N+](=O)[O-].Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65691MD View Documents Alfa Aesar™
H65691MD
250mg Each for $40.09
 
AAH6569103 View Documents Alfa Aesar™
H6569103
1g Each for $126.46
 
AAH6569106 View Documents Alfa Aesar™
H6569106
5g Each for $578.30
 
Specifications

Chemical Identifiers

40127-11-5
367.231
FBVMDLFQVOFFHS-XRIOVQLTSA-N
22811858
C1=CC(=CC=C1NC(=O)C(CCCN=C(N)N)N)[N+](=O)[O-].Cl.Cl
C12H20Cl2N6O3
MFCD00058249
h-arg-pna.2hcl, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide dihydrochloride, l-arginine p-nitroanilide dihydrochloride, arginine p-nitroanilide dihydrochloride, 2s-2-amino-5-carbamimidamido-n-4-nitrophenyl pentanamide dihydrochloride, h-arg-pna 2hcl, h-arg-pna.dihydrochloride, h-arg-pna inverted exclamation mark currency2hcl, n-4-nitrophenyl argininamide dihydrochloride, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide 2hcl
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-nitrophenyl)pentanamide;dihydrochloride

Specifications

40127-11-5
MFCD00058249
FBVMDLFQVOFFHS-XRIOVQLTSA-N
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-nitrophenyl)pentanamide;dihydrochloride
22811858
Solid
250mg
C12H20Cl2N6O3
h-arg-pna.2hcl, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide dihydrochloride, l-arginine p-nitroanilide dihydrochloride, arginine p-nitroanilide dihydrochloride, 2s-2-amino-5-carbamimidamido-n-4-nitrophenyl pentanamide dihydrochloride, h-arg-pna 2hcl, h-arg-pna.dihydrochloride, h-arg-pna inverted exclamation mark currency2hcl, n-4-nitrophenyl argininamide dihydrochloride, s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide 2hcl
C1=CC(=CC=C1NC(=O)C(CCCN=C(N)N)N)[N+](=O)[O-].Cl.Cl
367.231
367.23
95%
L-Arginine 4-nitroanilide dihydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C