missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(+)-Arabinose, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104980250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L(+)-Arabinose | |
155.0°C to 160.0°C | |
Authentic | |
Plastic bottle | |
25 g | |
10,783 | |
beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, pentopyranose #, b-arabinopyranose, beta-arabinopyranose 9ci, beta-arabinopyranose 9ci, a-d-arabinopyranoside, a-d-arabinopyranoside, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol | |
SRBFZHDQGSBBOR-VVZXFQNISA-N | |
(4S,5S)-oxane-2,3,4,5-tetrol | |
25245970 | |
99+% |
87-72-9 | |
White | |
99% min. (ELSD) (HPLC) | |
C5H10O5 | |
31,32 | |
+ 103.00 | |
(10% in water) Clear colorless | |
C1C(C(C(C(O1)O)O)O)O | |
150.13 | |
150.13 | |
Fine Crystalline Powder |
Chemical Identifiers
87-72-9 | |
150.13 | |
beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, pentopyranose #, b-arabinopyranose, beta-arabinopyranose 9ci, beta-arabinopyranose 9ci, a-d-arabinopyranoside, a-d-arabinopyranoside, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol | |
(4S,5S)-oxane-2,3,4,5-tetrol |
C5H10O5 | |
SRBFZHDQGSBBOR-VVZXFQNISA-N | |
25245970 | |
C1C(C(C(C(O1)O)O)O)O |
Safety and Handling
EINECSNumber : 201-767-6
RUO – Research Use Only