Please login to your online account to display your discounted pricing

L-AP6, Tocris


Selective agonist of the “quis” effect

Quantity 50mg

Manufacturer: r & d systems  RDS034150

 View more versions of this product

Catalog No. 034150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 211.15
Chemical Name or Material L-AP6
Assay Percent Range >98%
Molecular Formula C6H14NO5P
CAS 78944-89-5
Synonym L-(+)-2-Amino-6-phosphonohexanoic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material L-AP6
Assay Percent Range >98%
Molecular Formula C6H14NO5P
Formula Weight 211.15
Synonym L-(+)-2-Amino-6-phosphonohexanoic acid
Accessories

Store at Room Temperature