Please login to your online account to display your discounted pricing

L-AP6, Tocris

Selective agonist of the “quis” effect

$309.99 - $1,284.01

Chemical Identifiers

Chemical Name or Material L-AP6
Assay Percent Range >98%
Molecular Formula C6H14NO5P
Formula Weight 211.15
Synonym L-(+)-2-Amino-6-phosphonohexanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

034150

 
r & d systems
RDS034150
50mg Each for $1,284.01

034110

 
r & d systems
RDS034110
10mg Each for $309.99
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 211.15
Chemical Name or Material L-AP6
Assay Percent Range >98%
Molecular Formula C6H14NO5P
CAS 78944-89-5
Synonym L-(+)-2-Amino-6-phosphonohexanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature