Please login to your online account to display your discounted pricing

L-AP4, Tocris


Selective group III mGlu agonist

Quantity 10mg

Manufacturer: r & d systems  RDS010310

 View more versions of this product

Catalog No. 010310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 183.1
Assay Percent Range >99%
Molecular Formula C4H10NO5P
Chemical Name or Material L-AP4
Recommended Storage Store at Room Temperature
Synonym L-(+)-2-Amino-4-phosphonobutyric acid
CAS 23052-81-5

Specifications

Chemical Name or Material L-AP4
Assay Percent Range >99%
Molecular Formula C4H10NO5P
Formula Weight 183.1
Synonym L-(+)-2-Amino-4-phosphonobutyric acid
Accessories

Store at Room Temperature