Please login to your online account to display your discounted pricing

Alfa Aesar™ L-allo-Threonine, 99%

$294.00 - $772.72

Chemical Identifiers

Chemical Name or Material L-allo-Threonine
CAS 28954-12-3
Molecular Formula C4H9NO3
Formula Weight 119.12
MDL Number MFCD00064268
Beilstein 1721645
Synonym (2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2730103

 
alfa aesar™
H2730103
1g Each for $772.72

AAH27301MD

 
alfa aesar™
H27301MD
250mg Each for $294.00
Description & Specifications

Specifications

Formula Weight 119.12
Chemical Name or Material L-allo-Threonine
Assay 99%
Molecular Formula C4H9NO3
MDL Number MFCD00064268
CAS 28954-12-3
Beilstein 1721645
Synonym (2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH
Melting Point 272°C
Quantity 1g
Optical Rotation +9.2° (c=2 in Water)
RTECS Number BA4055000