missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alanyl-L-phenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H65182MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Alanyl-L-phenylalanine | |
C12H16N2O3 | |
2381598 | |
OMNVYXHOSHNURL-WPRPVWTQSA-N | |
(2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-phenylpropanoic acid | |
96814 | |
236.27 | |
250 mg |
3061-90-3 | |
MFCD00066031 | |
ala-phe, h-ala-phe-oh, l-alanyl-l-phenylalanine, alanylphenylalanine, s-2-s-2-aminopropanamido-3-phenylpropanoic acid, l-ala-l-phe, chembl57338, ala-phe-oh, phenylalanine, alanyl, 2-2-amino-propionylamino-3-phenyl-propionic acid | |
CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N | |
236.271 | |
CHEBI:73807 | |
95% | |
Solid |
Chemical Identifiers
3061-90-3 | |
236.271 | |
OMNVYXHOSHNURL-WPRPVWTQSA-N | |
96814 | |
(2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-phenylpropanoic acid |
C12H16N2O3 | |
MFCD00066031 | |
ala-phe, h-ala-phe-oh, l-alanyl-l-phenylalanine, alanylphenylalanine, s-2-s-2-aminopropanamido-3-phenylpropanoic acid, l-ala-l-phe, chembl57338, ala-phe-oh, phenylalanine, alanyl, 2-2-amino-propionylamino-3-phenyl-propionic acid | |
CHEBI:73807 | |
CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only