Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Alanyl-L-glutamine, 99%


Quantity 100g

Manufacturer: alfa aesar™  J6699622

 View more versions of this product

Catalog No. AAJ6699622

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 217.22
CAS 39537-23-0
Chemical Name or Material L-Alanyl-L-glutamine
Physical Form Powder
MDL Number MFCD00133046
Synonym Ala-Gln; Glutamine-S
Assay 99%
Solubility Information Soluble in water
Boiling Point 615°C
Color White
Quantity 100g
Storage Note 1 2° to 8°C
RTECS Number MA2275262
Melting Point 222°C

Specifications

Chemical Name or Material L-Alanyl-L-glutamine
CAS 39537-23-0
Formula Weight 217.22
MDL Number MFCD00133046
Synonym Ala-Gln; Glutamine-S

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Store in a cool place. Incompatible with strong oxidizing agents.

Accessories

2° to 8°C

SDS & Safety Documents