missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alanine tert-butyl ester hydrochloride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 442920010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L-Alanine tert-butyl ester hydrochloride | |
99% | |
MFCD00035524 | |
h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride | |
Cl.CC(N)C(=O)OC(C)(C)C | |
181.66 | |
181.66 |
13404-22-3 | |
C7H16ClNO2 | |
Glass bottle | |
WIQIWPPQGWGVHD-UHFFFAOYNA-N | |
tert-butyl (2S)-2-aminopropanoate;hydrochloride | |
11275390 | |
1g |
Safety and Handling
EINECSNumber : 236-497-8
RUO â Research Use Only