missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Alanine isopropyl ester hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H62490MD

 View more versions of this product

Catalog No. AAH62490MD



Description

Description

Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

L-Alanine isopropyl ester hydrochloride
C6H13NO2
s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
CC(C)OC(=O)C(C)N
131.175
167.63
250mg
39825-33-7
MFCD08059709
QDQVXVRZVCTVHE-YFKPBYRVSA-N
propan-2-yl (2S)-2-aminopropanoate
13089460
95%

Chemical Identifiers

39825-33-7
131.175
QDQVXVRZVCTVHE-YFKPBYRVSA-N
13089460
CC(C)OC(=O)C(C)N
C6H13NO2
MFCD08059709
s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
propan-2-yl (2S)-2-aminopropanoate
SDS Documents