missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Alanine ethyl ester hydrochloride, 98+%

Manufacturer:  Alfa Aesar™ A1127106

 View more versions of this product

Catalog No. AAA1127106


 

Specifications

Specifications

L-Alanine ethyl ester hydrochloride
≥98%
MFCD00063662
H-Ala-OEt.HCl
CCOC(=O)C(C)N.Cl
153.606
153.61
5g
1115-59-9
C5H12ClNO2
3594395
JCXLZWMDXJFOOI-WCCKRBBISA-N
ethyl (2S)-2-aminopropanoate;hydrochloride
2724356
74°C to 78°C
+2.9° (c=2.5 in water)

Chemical Identifiers

1115-59-9
153.606
JCXLZWMDXJFOOI-WCCKRBBISA-N
2724356
C5H12ClNO2
MFCD00063662
H-Ala-OEt.HCl
SDS Documents