Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Alanine benzyl ester p-toluenesulfonate, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1074306

 View more versions of this product

Catalog No. AAL1074306

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 351.42
Chemical Name or Material L-Alanine benzyl ester p-toluenesulfonate
Assay 98%
Molecular Formula C17H21NO5S
Beilstein 3586474
Synonym H-Ala-OBzl.TosOH
Melting Point 114° to 116°C
Quantity 5g
MDL Number MFCD00066143
CAS 42854-62-6
Optical Rotation −5.3° (c=3 in Methanol)
Recommended Storage 2° to 8°C

Specifications

Chemical Name or Material L-Alanine benzyl ester p-toluenesulfonate
CAS 42854-62-6
Molecular Formula C17H21NO5S
Formula Weight 351.42
MDL Number MFCD00066143
Beilstein 3586474
Synonym H-Ala-OBzl.TosOH
Accessories

2° to 8°C