missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Alanine benzyl ester hydrochloride, 98%

$40.72 - $527.41

Chemical Identifiers

CAS 5557-83-5
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00054340
InChI Key RLMHWGDKMJIEHH-QRPNPIFTSA-N
Synonym h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
PubChem CID 12210893
IUPAC Name benzyl (2S)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OCC1=CC=CC=C1)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6569906 View Documents Alfa Aesar™
H6569906
5g Each for $40.72
 
AAH6569914 View Documents Alfa Aesar™
H6569914
25g Each for $163.46
 
AAH6569922 View Documents Alfa Aesar™
H6569922
100g Each for $527.41
 
Specifications

Chemical Identifiers

5557-83-5
215.677
RLMHWGDKMJIEHH-QRPNPIFTSA-N
12210893
CC(C(=O)OCC1=CC=CC=C1)N.Cl
C10H14ClNO2
MFCD00054340
h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
benzyl (2S)-2-aminopropanoate;hydrochloride

Specifications

5557-83-5
MFCD00054340
RLMHWGDKMJIEHH-QRPNPIFTSA-N
benzyl (2S)-2-aminopropanoate;hydrochloride
12210893
Powder
White
L-Alanine benzyl ester hydrochloride
C10H14ClNO2
h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
CC(C(=O)OCC1=CC=CC=C1)N.Cl
215.677
215.68
98%
5g
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C