missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H63393MD

 View more versions of this product

Catalog No. AAH63393MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Alanine 4-nitroanilide hydrochloride
Yellow
MFCD00039088
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl
245.663
245.66
250 mg
31796-55-1
C9H12ClN3O3
6303731
YEXRLSXNWLNHQR-RGMNGODLSA-N
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride
2802426
98%
Powder

Chemical Identifiers

31796-55-1
245.663
YEXRLSXNWLNHQR-RGMNGODLSA-N
2802426
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl
C9H12ClN3O3
MFCD00039088
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : 250-811-0

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only