missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alaninamide hydrochloride, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442950050
Specifications
L-Alaninamide hydrochloride | |
213°C to 215°C | |
33208-99-0 | |
C3H8N2O·ClH | |
FIAINKIUSZGVGX-DKWTVANSSA-N | |
(2S)-2-aminopropanamide;hydrochloride | |
2775816 | |
95% |
11.5 | |
5g | |
94 to max. 106% min. (Argentometry) | |
2s-2-aminopropanamide hydrochloride, ala-nh2 hcl, alaninamide, hydrochloride, h-ala-nh2.hcl, l-alaninamide hcl, l-alaninamide hydrochloride, l-alaninamidehydrochloride, l-alanine amide hydrochloride, s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride | |
CC(C(=O)N)N.Cl | |
124.57 | |
124.57 |
Chemical Identifiers
33208-99-0 | |
124.57 | |
2s-2-aminopropanamide hydrochloride, ala-nh2 hcl, alaninamide, hydrochloride, h-ala-nh2.hcl, l-alaninamide hcl, l-alaninamide hydrochloride, l-alaninamidehydrochloride, l-alanine amide hydrochloride, s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride | |
(2S)-2-aminopropanamide;hydrochloride |
C3H8N2O·ClH | |
FIAINKIUSZGVGX-DKWTVANSSA-N | |
2775816 | |
CC(C(=O)N)N.Cl |