missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alaninamide hydrochloride, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 33208-99-0 |
---|---|
Molecular Formula | C3H8N2O·ClH |
Molecular Weight (g/mol) | 124.57 |
InChI Key | FIAINKIUSZGVGX-DKWTVANSSA-N |
Synonym | l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride |
PubChem CID | 2775816 |
IUPAC Name | (2S)-2-aminopropanamide;hydrochloride |
SMILES | CC(C(=O)N)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC442950010
|
Thermo Scientific Chemicals
442950010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC442950050
|
Thermo Scientific Chemicals
442950050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
33208-99-0 | |
124.57 | |
l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride | |
(2S)-2-aminopropanamide;hydrochloride |
C3H8N2O·ClH | |
FIAINKIUSZGVGX-DKWTVANSSA-N | |
2775816 | |
CC(C(=O)N)N.Cl |
Specifications
33208-99-0 | |
Glass Bottle | |
1g | |
l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride | |
CC(C(=O)N)N.Cl | |
124.57 | |
124.57 | |
L-Alaninamide hydrochloride |
94 to max. 106% min. (Argentometry) | |
C3H8N2O·ClH | |
11.5 | |
FIAINKIUSZGVGX-DKWTVANSSA-N | |
(2S)-2-aminopropanamide;hydrochloride | |
2775816 | |
95% |
RUO â Research Use Only