missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Adrenaline 98.0+%, TCI America™

$244.45 - $733.34

Chemical Identifiers

CAS 51-43-4
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00002204
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
Synonym epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
PubChem CID 5816
ChEBI CHEBI:28918
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A01735G View Documents TCI America
A01735G
5g Each for $244.45
 
A017325G View Documents TCI America
A017325G
25g Each for $733.34
 
Specifications

Chemical Identifiers

51-43-4
183.207
UCTWMZQNUQWSLP-VIFPVBQESA-N
5816
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
C9H13NO3
MFCD00002204
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
CHEBI:28918
CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

White-Yellow
5g
C9H13NO3
2811
UCTWMZQNUQWSLP-VIFPVBQESA-N
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
5816
183.21
≥98.0% (T)
212°C
51-43-4
MFCD00002204
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
CNCC(C1=CC(=C(C=C1)O)O)O
183.207
CHEBI:28918
Crystal-Powder at 20°C
L-Adrenaline
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0049

RTECSNumber : DO2625000

TSCA : Yes

Recommended Storage : Refrigerator