Please login to your online account to display your discounted pricing

L-798,106, Tocris

Potent and highly selective EP3 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material L-798, 106
Assay Percent Range >99%
Molecular Formula C27H22BrNO4S
Formula Weight 536.44
Synonym N-[(5-Bromo-2-methoxyphenyl)sulfonyl]-3-[2-(2-naphthalenylmethyl)phenyl]-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

334250

 
r & d systems
RDS334250
50mg Each for $1,050.42

334210

 
r & d systems
RDS334210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 536.44
Quantity 50mg
Recommended Storage Store at Room Temperature
Chemical Name or Material L-798, 106
Assay Percent Range >99%
Molecular Formula C27H22BrNO4S
CAS 244101-02-8
Synonym N-[(5-Bromo-2-methoxyphenyl)sulfonyl]-3-[2-(2-naphthalenylmethyl)phenyl]-2-propenamide
Safety and Handling

Store at Room Temperature