Please login to your online account to display your discounted pricing

L-779,450, Tocris

Potent Raf kinase inhibitor

$241.15 - $1,015.48

Chemical Identifiers

Chemical Name or Material L-779, 450
Assay Percent Range >99%
Molecular Formula C20H14ClN3O
Formula Weight 347.8
Synonym 2-Chloro-5-[2-Phenyl-5-(4-pyridinyl)-1H-imidazol-4-yl]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

318510

 
r & d systems
RDS318510
10mg Each for $241.15

318550

 
r & d systems
RDS318550
50mg Each for $1,015.48
Description & Specifications

Specifications

Formula Weight 347.8
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C20H14ClN3O
Synonym 2-Chloro-5-[2-Phenyl-5-(4-pyridinyl)-1H-imidazol-4-yl]phenol
Recommended Storage Store at +4°C
Chemical Name or Material L-779, 450
CAS 303727-31-3
Safety and Handling

Store at +4°C