Please login to your online account to display your discounted pricing

L-745,870 trihydrochloride, Tocris


Highly selective D4 antagonist

Quantity 10mg

Manufacturer: r & d systems  100210

 View more versions of this product

Catalog No. 100210

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material L-745, 870 trihydrochloride
Quantity 10mg
CAS 866021-03-6
Recommended Storage Desiccate at +4°C
Assay Percent Range >99%
Molecular Formula C18H19N4Cl·3HCl
Formula Weight 436.21
Synonym 3-(4-[4-Chlorophenyl]piperazin-1-yl)-methyl-1H-pyrrolo[2, 3-b]pyridine trihydrochloride

Specifications

Chemical Name or Material L-745, 870 trihydrochloride
Assay Percent Range >99%
Molecular Formula C18H19N4Cl·3HCl
Formula Weight 436.21
Synonym 3-(4-[4-Chlorophenyl]piperazin-1-yl)-methyl-1H-pyrrolo[2, 3-b]pyridine trihydrochloride
Accessories

Desiccate at +4°C