Please login to your online account to display your discounted pricing

L-733,060 hydrochloride, Tocris

Potent NK1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material L-733, 060 hydrochloride
Assay Percent Range >99%
Molecular Formula C20H19F6NO·HCl
Formula Weight 439.83
Synonym (2S, 3S)-3-[[3, 5-bis(Trifluoromethyl)phenyl]methoxy]-2-phenylpiperidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

114510

 
r & d systems
RDS114510
10mg Each for $252.39

114550

 
r & d systems
RDS114550
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 439.83
Quantity 10mg
CAS 148687-76-7
Molecular Formula C20H19F6NO·HCl
Assay Percent Range >99%
Chemical Name or Material L-733, 060 hydrochloride
Synonym (2S, 3S)-3-[[3, 5-bis(Trifluoromethyl)phenyl]methoxy]-2-phenylpiperidine hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature