Please login to your online account to display your discounted pricing

L-732,138, Tocris


Potent, selective NK1 antagonist

Quantity 10mg

Manufacturer: r & d systems  086810

 View more versions of this product

Catalog No. 086810

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material L-732, 138
CAS 148451-96-1
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C20H18F6N2O3
Formula Weight 472.39
Synonym N-Acetyl-L-tryptophan 3, 5-bis(trifluoromethyl)benzyl ester
Quantity 10mg

Specifications

Chemical Name or Material L-732, 138
Assay Percent Range >98%
Molecular Formula C20H18F6N2O3
Formula Weight 472.39
Synonym N-Acetyl-L-tryptophan 3, 5-bis(trifluoromethyl)benzyl ester
Accessories

Store at Room Temperature