Please login to your online account to display your discounted pricing

L-732,138, Tocris

Potent, selective NK1 antagonist

$113.57 - $437.93

Chemical Identifiers

Chemical Name or Material L-732, 138
Assay Percent Range >98%
Molecular Formula C20H18F6N2O3
Formula Weight 472.39
Synonym N-Acetyl-L-tryptophan 3, 5-bis(trifluoromethyl)benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

086810

 
r & d systems
086810
10mg Each for $113.57

086850

 
r & d systems
086850
50mg Each for $437.93
Description & Specifications

Specifications

Chemical Name or Material L-732, 138
CAS 148451-96-1
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C20H18F6N2O3
Formula Weight 472.39
Synonym N-Acetyl-L-tryptophan 3, 5-bis(trifluoromethyl)benzyl ester
Quantity 10mg
Safety and Handling

Store at Room Temperature