Please login to your online account to display your discounted pricing

L-703,664 succinate, Tocris

Selective 5-HT1D receptor agonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material L-703, 664 succinate
Assay Percent Range >99%
Molecular Formula C16H24N4O2S·C4H6O4
Formula Weight 454.54
Synonym N,N-Dimethyl-5-[(5-methyl-1,1-dioxodo-1,2,5-thiadiazolidin-2-yl)methyl]-1H-indole-3-ethanamine succinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264010

 
r & d systems
RDS264010
10mg Each for $257.85

264050

 
r & d systems
RDS264050
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 454.54
Quantity 10mg
Chemical Name or Material L-703, 664 succinate
Assay Percent Range >99%
CAS 144776-01-2
Synonym N,N-Dimethyl-5-[(5-methyl-1,1-dioxodo-1,2,5-thiadiazolidin-2-yl)methyl]-1H-indole-3-ethanamine succinate
Molecular Formula C16H24N4O2S·C4H6O4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C