Please login to your online account to display your discounted pricing

L-701,324, Tocris

NMDA antagonist, acts at glycine site

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material L-701, 324
Assay Percent Range >99%
Molecular Formula C21H14ClNO3
Formula Weight 363.8
Synonym 7-Chloro-4-hydroxy-3-(3-phenoxy)phenyl-2(1H)-quinolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

090710

 
r & d systems
RDS090710
10mg Each for $181.19

090750

 
r & d systems
RDS090750
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 363.8
Chemical Name or Material L-701, 324
Assay Percent Range >99%
Molecular Formula C21H14ClNO3
CAS 142326-59-8
Synonym 7-Chloro-4-hydroxy-3-(3-phenoxy)phenyl-2(1H)-quinolinone
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature