Please login to your online account to display your discounted pricing

L-701,252, Tocris


NMDA antagonist, acts glycine site

Quantity 10mg

Manufacturer: r & d systems  RDS070510

 View more versions of this product

Catalog No. 070510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 263.68
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
CAS 151057-13-5
Chemical Name or Material L-701, 252
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material L-701, 252
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
Formula Weight 263.68
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
Accessories

Desiccate at +4°C