Please login to your online account to display your discounted pricing

L-701,252, Tocris

NMDA antagonist, acts glycine site

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material L-701, 252
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
Formula Weight 263.68
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

070510

 
r & d systems
RDS070510
10mg Each for $181.19

070550

 
r & d systems
RDS070550
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 263.68
Chemical Name or Material L-701, 252
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
CAS 151057-13-5
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C