Please login to your online account to display your discounted pricing

L-701,252, Tocris


NMDA antagonist, acts glycine site

Quantity 50mg

Manufacturer: r & d systems  RDS070550

 View more versions of this product

Catalog No. 070550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 263.68
Chemical Name or Material L-701, 252
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
CAS 151057-13-5
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material L-701, 252
Assay Percent Range >99%
Molecular Formula C13H10ClNO3
Formula Weight 263.68
Synonym 7-Chloro-3-(cyclopropylcarbonyl)-4-hydroxy-2(1H)-quinolinone
Accessories

Desiccate at +4°C