Please login to your online account to display your discounted pricing

L-693,403 maleate, Tocris


High affinity σ ligand

Quantity 10mg

Manufacturer: r & d systems  042610

 View more versions of this product

Catalog No. 042610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 393.48
Assay Percent Range >98%
Recommended Storage Desiccate at -20°C
Synonym N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate
Molecular Formula C20H23N·C4H4O4
CAS 207455-21-8
Chemical Name or Material L-693, 403 maleate

Specifications

Chemical Name or Material L-693, 403 maleate
Assay Percent Range >98%
Molecular Formula C20H23N·C4H4O4
Formula Weight 393.48
Synonym N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate
Accessories

Desiccate at -20°C