Please login to your online account to display your discounted pricing

L-693,403 maleate, Tocris

High affinity σ ligand

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material L-693, 403 maleate
Assay Percent Range >98%
Molecular Formula C20H23N·C4H4O4
Formula Weight 393.48
Synonym N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

042610

 
r & d systems
042610
10mg Each for $195.49

042650

 
r & d systems
042650
50mg Each for $816.90
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 393.48
Assay Percent Range >98%
Recommended Storage Desiccate at -20°C
Synonym N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate
Molecular Formula C20H23N·C4H4O4
CAS 207455-21-8
Chemical Name or Material L-693, 403 maleate
Safety and Handling

Desiccate at -20°C