missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-693,403 maleate, Tocris

High affinity σ ligand

Manufacturer:  R & D Systems 042650

 View more versions of this product

Catalog No. 042650


 

Specifications Description & Specifications

Specifications

L-693, 403 maleate
>98%
C20H23N·C4H4O4
N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate
207455-21-8
50mg
393.48

Chemical Identifiers

>98%
393.48
L-693, 403 maleate
C20H23N·C4H4O4
N-1'-Benzyl-3, 4-dihydrospiro[1H-indene-1, 4'-piperidine] maleate

Safety and Handling

Recommended Storage : Desiccate at -20°C