Please login to your online account to display your discounted pricing

L-655,240, Tocris

Potent, selective thromboxane A2/prostaglandin endoperoxide antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material L-655, 240
Assay Percent Range >99%
Molecular Formula C21H21ClFNO2
Formula Weight 373.85
Synonym 1-[(4-Chlorophenyl)methyl]-5-fluoro-α, α, 3-trimethyl-1H-indole-2-propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

169810

 
r & d systems
169810
10mg Each for $201.36

169850

 
r & d systems
169850
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 373.85
Quantity 10mg
Chemical Name or Material L-655, 240
Assay Percent Range >99%
Molecular Formula C21H21ClFNO2
CAS 103253-15-2
Synonym 1-[(4-Chlorophenyl)methyl]-5-fluoro-α, α, 3-trimethyl-1H-indole-2-propanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature