Please login to your online account to display your discounted pricing

L-4-chlorophenylalanine, 98%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  302941000

 View more versions of this product

Catalog No. AC302941000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 199.64
Quantity 100mg
Color White
Linear Formula ClC6H4CH2CH(NH2)CO2H
Packaging Glass bottle
Chemical Name or Material L-4-Chlorophenylalanine
Assay Percent Range 98%
Molecular Formula C9H10ClNO2
CAS 14173-39-8
Specific Rotation -23° to -27° (24°C, 589nm) (c=0.5, H2O)
MDL Number MFCD00063065
Infrared Spectrum Authentic

Specifications

Chemical Name or Material L-4-Chlorophenylalanine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10ClNO2
Linear Formula ClC6H4CH2CH(NH2)CO2H
Formula Weight 199.64
MDL Number MFCD00063065