Please login to your online account to display your discounted pricing

L-4-chlorophenylalanine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-4-Chlorophenylalanine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10ClNO2
Linear Formula ClC6H4CH2CH(NH2)CO2H
Formula Weight 199.64
MDL Number MFCD00063065
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC302941000

  SDS
thermo fisher scientific
302941000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 199.64
Physical Form Powder
Quantity 100mg
Color White
Packaging Glass bottle
Linear Formula ClC6H4CH2CH(NH2)CO2H
Chemical Name or Material L-4-Chlorophenylalanine
CAS 14173-39-8
Molecular Formula C9H10ClNO2
Assay Percent Range 98%
MDL Number MFCD00063065
Infrared Spectrum Authentic
Specific Rotation -23° to -27° (24°C, 589nm) (c=0.5, H2O)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.