Please login to your online account to display your discounted pricing

L-368,899 hydrochloride, Tocris

Potent, non-peptide oxytocin receptor antagonist

$172.26 - $368.77

Chemical Identifiers

Chemical Name or Material L-368, 899 hydrochloride
Assay Percent Range >97%
Molecular Formula C26H42N4O5S2·HCl
Formula Weight 591.23
Synonym (2S)-2-Amino-N-[(1S,2S,4R)-7,7-dimethyl-1-[[[4-(2-methylphenyl)-1-piperazinyl]sulfonyl]methyl]bicyclo[2.2.1]hept-2-yl]-4-(methylsulfonyl)butanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264110

 
r & d systems
RDS264110
10mg Each for $368.77

26411

 
r & d systems
RDS26411
1mg Each for $172.26
Description & Specifications

Specifications

Formula Weight 591.23
Quantity 10mg
Chemical Name or Material L-368, 899 hydrochloride
Assay Percent Range >97%
Molecular Formula C26H42N4O5S2·HCl
CAS 160312-62-9
Synonym (2S)-2-Amino-N-[(1S,2S,4R)-7,7-dimethyl-1-[[[4-(2-methylphenyl)-1-piperazinyl]sulfonyl]methyl]bicyclo[2.2.1]hept-2-yl]-4-(methylsulfonyl)butanamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C