Please login to your online account to display your discounted pricing

L-364,373, Tocris

KV7.1 channel activator, activates Iks

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material L-364, 373
Assay Percent Range >98%
Molecular Formula C25H20FN3O
Formula Weight 397.44
Synonym 5-(2-Fluorophenyl)-1, 3-dihydro-3-(1H-indol-3-ylmethyl)-1-methyl-2H-1, 4-benzodiazepin-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

266050

 
r & d systems
266050
50mg Each for $1,073.10

266010

 
r & d systems
266010
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 397.44
Quantity 50mg
Chemical Name or Material L-364, 373
Assay Percent Range >98%
Molecular Formula C25H20FN3O
CAS 103342-82-1
Synonym 5-(2-Fluorophenyl)-1, 3-dihydro-3-(1H-indol-3-ylmethyl)-1-methyl-2H-1, 4-benzodiazepin-2-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C