missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(-)-3-Phenyllactic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 150920050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L(-)-3-Phenyllactic acid | |
Authentic | |
Glass bottle | |
C6H5CH2CH(OH)CO2H | |
5g | |
-18.5 | |
Solubility in water: soluble | |
O[C@@H](CC1=CC=CC=C1)C([O-])=O | |
165.17 | |
CHEBI:43065 | |
98% |
20312-36-1 | |
98% | |
C9H9O3 | |
MFCD00004244 | |
−18.50° (24°C c=1,CH3COOH) | |
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
(2S)-2-hydroxy-3-phenylpropanoic acid | |
444718 | |
166.18 |
Safety and Handling
EINECSNumber : 243-726-5
RUO â Research Use Only