missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

$263.33 - $263.33

Chemical Identifiers

CAS 1421-65-4
Molecular Formula C10H14ClNO4
Molecular Weight (g/mol) 247.68
InChI Key WFGNJLMSYIJWII-UHFFFAOYNA-N
Synonym s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105
PubChem CID 10131132
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439490010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
439490010
1 g
Each for $263.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1421-65-4
247.68
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105
[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
C10H14ClNO4
WFGNJLMSYIJWII-UHFFFAOYNA-N
10131132

Specifications

1421-65-4
White
Authentic
Glass Bottle
1 g
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride, l-3,4-dihydroxyphenylalanine methyl ester hydrochloride, methyl l-dopa hydrochloride, melevodopa hydrochloride, unii-jfu1a8866v, l-dopa methyl ester hydrochloride, levodopa methyl ester hydrochloride, l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride, dsstox_cid_25763, dsstox_rid_81105
[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
247.68
247.68
Crystalline Powder or Powder
94.2°C to 94.3°C
45.58 to 51.40 % (C) Dry basis 5.32 to 5.99 % (N) Dry basis
95%
C10H14ClNO4
+9.0° to +11.0° (20°C, 589nm) (C=2 in methanol)
WFGNJLMSYIJWII-UHFFFAOYNA-N
methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride
10131132
95%
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only