missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-2-Phenylglycine methyl ester hydrochloride, 97%

$57.82 - $206.65

Chemical Identifiers

CAS 15028-39-4
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00077158
InChI Key DTHMTBUWTGVEFG-QRPNPIFTSA-N
Synonym h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
PubChem CID 11217910
IUPAC Name methyl (2S)-2-amino-2-phenylacetate;hydrochloride
SMILES COC(=O)C(C1=CC=CC=C1)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6287414 View Documents Alfa Aesar™
H6287414
25g Each for $57.82
 
AAH6287422 View Documents Alfa Aesar™
H6287422
100g Each for $206.65
 
Specifications

Chemical Identifiers

15028-39-4
201.65
DTHMTBUWTGVEFG-QRPNPIFTSA-N
11217910
COC(=O)C(C1=CC=CC=C1)N.Cl
C9H12ClNO2
MFCD00077158
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
methyl (2S)-2-amino-2-phenylacetate;hydrochloride

Specifications

15028-39-4
MFCD00077158
DTHMTBUWTGVEFG-QRPNPIFTSA-N
methyl (2S)-2-amino-2-phenylacetate;hydrochloride
11217910
97%
25g
L-(+)-2-Phenylglycine methyl ester hydrochloride
C9H12ClNO2
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
COC(=O)C(C1=CC=CC=C1)N.Cl
201.65
201.65
200°C
+118° (c=1 in Water)
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning