missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6287422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
200°C (decomposition) | |
15028-39-4 | |
MFCD00077158 | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride | |
11217910 | |
97% | |
+118° (c=1 in Water) |
L-(+)-2-Phenylglycine methyl ester hydrochloride | |
C9H12ClNO2 | |
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride | |
COC(=O)C(C1=CC=CC=C1)N.Cl | |
201.65 | |
201.65 | |
100 g |
Chemical Identifiers
15028-39-4 | |
201.65 | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
11217910 | |
COC(=O)C(C1=CC=CC=C1)N.Cl |
C9H12ClNO2 | |
MFCD00077158 | |
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only