missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-2-Octanol, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164590100

 View more versions of this product

Catalog No. AC164590100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L(-)-2-Octanol
60°C
Glass bottle
0.83
− 9.50
Colorless to Yellow
99+%
99% min. (GC)
CH3(CH2)5CH(OH)CH3
01, IV, 1770
2r-2-octanol
SJWFXCIHNDVPSH-MRVPVSSYSA-N
(2R)-octan-2-ol
80080
130.23
99+%
0.8300g/mL
Authentic
1.4250 to 1.4270
−9.50° (20°C neat)
175.0°C
10g
5978-70-1
C8H18O
MFCD00064284
15, 6841
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether
CCCCCCC(C)O
130.23
CHEBI:37871
Liquid

Chemical Identifiers

5978-70-1
130.23
SJWFXCIHNDVPSH-MRVPVSSYSA-N
80080
(2R)-octan-2-ol
C8H18O
MFCD00064284
2r-2-octanol
CHEBI:37871
CCCCCCC(C)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 227-777-0

TSCA : TSCA

SDS Documents