missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-(+)-2-Cyclohexylglycine, 98%

$182.50 - $543.01

Chemical Identifiers

CAS 14328-51-9
Molecular Formula C8H15NO2
Formula Weight 157.22
MDL Number MFCD01311679
Synonym (S)-(-)-alpha-Aminocyclohexaneacetic acid; H-Cyclohexyl-Gly-OH
Chemical Name or Material L-(+)-2-Cyclohexylglycine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2688103 SDS Alfa Aesar™
H2688103
1g Each for $182.50
AAH2688106 SDS Alfa Aesar™
H2688106
5g Each for $543.01
Specifications Description & Specifications

Chemical Identifiers

14328-51-9
157.22
(S)-(-)-alpha-Aminocyclohexaneacetic acid; H-Cyclohexyl-Gly-OH
C8H15NO2
MFCD01311679
L-(+)-2-Cyclohexylglycine

Specifications

14328-51-9
157.22
(S)-(-)-alpha-Aminocyclohexaneacetic acid; H-Cyclohexyl-Gly-OH
1g
L-(+)-2-Cyclohexylglycine
C8H15NO2
MFCD01311679
>300°C
+31° (c=0.5 in 1N HCl)
98%