Please login to your online account to display your discounted pricing

L(+)-2-Aminobutyric acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  215380050

 View more versions of this product

Catalog No. AC215380050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 103.12
Quantity 5g
Color White to beige
Beilstein 04, 408
Merck Index 10, 430
EINECS Number 216-083-3
TSCA TSCA
Chemical Name or Material L(+)-2-Aminobutyric acid
Packaging Glass bottle
Sulfated Ash 0.1% max.
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H9NO2
Linear Formula C2H5CH(NH2)CO2H
MDL Number MFCD00064415
CAS 1492-24-6
Synonym (S)-(+)-2-Aminobutyric acid
Specific Rotation +20.2° to +23° (20°C, 589nm) (c=2, 5N HCl)
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material L(+)-2-Aminobutyric acid
Name Note 98%
CAS 1492-24-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H9NO2
Linear Formula C2H5CH(NH2)CO2H
Formula Weight 103.12
MDL Number MFCD00064415
Synonym (S)-(+)-2-Aminobutyric acid