Please login to your online account to display your discounted pricing

L-2-Aminoadipic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC302920010

 View more versions of this product

Catalog No. AC302920010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Solid
Formula Weight 161.16
Quantity 1g
Color White
Specific Rotation Condition +22° (25°C c=2,5 NHCl)
Packaging Glass bottle
MDL Number MFCD00002636
Infrared Spectrum Authentic
Chemical Name or Material L-2-Aminoadipic acid
Name Note 98%
Molecular Formula C6H11NO4
Linear Formula HOC(CH)3CH(NH)COH
CAS 1118-90-7
Specific Rotation +20° to +24° (25°C, 589nm) (c=2, 5N HCl)
Assay Percent Range 98%

Specifications

Chemical Name or Material L-2-Aminoadipic acid
Name Note 98%
CAS 1118-90-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11NO4
Linear Formula HOC(CH)3CH(NH)COH
Formula Weight 161.16
MDL Number MFCD00002636